Learn More
ADK Mouse anti-Human, Unconjugated, Clone: 1G1A5, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66929-1-IG-20UL
This item is not returnable.
View return policy
Description
The ADK antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ADK. This antibody recognizes human, mouse, rat antigen. The ADK antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
ADK | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human, Rat, Mouse |
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
1G1A5 | |
Unconjugated | |
20 μL | |
132 | |
IgG1 |