missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADK Mouse anti-Human, Unconjugated, Clone: 1G1A5, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
Antigen | ADK |
---|---|
Clone | 1G1A5 |
Concentration | 1000 μg/mL |
Applications | Western Blot, Immunohistochemistry, Immunofluorescence, ELISA |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16829235
|
Proteintech
66929-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16819235
|
Proteintech
66929-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The ADK antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ADK. This antibody recognizes human, mouse, rat antigen. The ADK antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
ADK | |
1000 μg/mL | |
Monoclonal | |
Mouse | |
132 | |
Primary |
1G1A5 | |
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
Unconjugated | |
Human, Rat, Mouse | |
IgG1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title