Learn More
Advanced Instruments Osmometer Standards
For in vitro diagnostic use
Brand:  Advanced Instruments 3MA040
Encompass
 View more versions of this product
Product Code. 13284750
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
For in vitro diagnostic use
  • 2-Hydroxybiphenyl sodium salt tetrahydrate
  • 1% added as preservative
Specifications
Specifications
2°C to 30°C
3250, 3900 and A2O Osmometers
2- Hydroxybiphenyl sodium salt tetrahydrate, 0.01% added as preservative
10 x 2 mL
400 mOsm/kg H2O
Osmometer Calibration Standard
To check the performance of your osmometer and test methods, in-vitro diagnostic use