Learn More
METTLER TOLEDO Advanced MX Analytical Balance
METTLER TOLEDO Advanced MX Analytical Balances offer a brand new feature, the Backlit Draft Shield (5-place models only). Brightness extends over the draft shield's entire back panel providing exceptional visibility of the work area for highly precise dosing.
Brand:  METTLER TOLEDO 30665097
 View more versions of this product
Product Code. 18032284
  • £11290.00 / Each
Explore more special offers
Description
Show More
Specifications
Specifications
USB-A, USB-C, Ethernet
Color Touchscreen
Metal
2 sec.
100/240 V, 50/60 Hz
120 g
0.01 mg
MX105
Analytical Balance
No
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
METTLER TOLEDO Advanced MX Analytical Balance > Model: MX105; Capacity: 120 g; Readability: 0.01 mg