Learn More
Ratiolab™ Aeroject™ Ultra Filter Tips
Pipette tips with special filters to effectively prevent the pipettor shaft from contamination with aerosols and liquids to be sucked into the pipettor.
Brand: Ratiolab™ 1771000
Code : NEW
Additional Details : Weight : 3.00000kg
Description
- Sterilized
- Free of RNase-, DNase-, DNA-, pyrogens, ATP
- Free of human DNA and PCR inhibitors
- Pipet tips and aerosol filters made of ultra-hydrophobic PE-HD
- Minimized retention due to special surface treatment
- Perfect and tight fit on all major pipettor brands
- Packed in plastic racks with hinged lids
Specifications
1000 μL | |
Sterile | |
70.10 mm | |
Hinged Rack | |
600 tips | |
Aeroject Ultra Filter |
Filtered | |
Blue | |
Contamination prevention | |
Low Retention | |
Universal Fit |