missing translation for 'onlineSavingsMsg'
Learn More

Ratiolab™ Aeroject™ Ultra Filter Tips

Pipette tips with special filters to effectively prevent the pipettor shaft from contamination with aerosols and liquids to be sucked into the pipettor.

£59.04 - £83.85

Specifications

Filtered Filtered
Sterility Sterile
Rack Color Blue
For Use With (Application) Contamination prevention
Format Hinged Rack
View More Specs

Products 2
Product Code Brand Volume (Metric) Quantity Price Quantity & Availability  
Product Code Brand Volume (Metric) Quantity Price Quantity & Availability  
15370955
View Documents Promotion Details
Ratiolab™
1770100
100 μL 960 tips
£83.85
Pack of 960
View alternative products
Save Up to 
»
Estimated Shipment: 30-12-2024
to see stock.
Add to basket
 
15300965
View Documents Promotion Details
Ratiolab™
1771000
1000 μL 600 tips This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

  • Sterilized
  • Free of RNase-, DNase-, DNA-, pyrogens, ATP
  • Free of human DNA and PCR inhibitors
  • Pipet tips and aerosol filters made of ultra-hydrophobic PE-HD
  • Minimized retention due to special surface treatment
  • Perfect and tight fit on all major pipettor brands
  • Packed in plastic racks with hinged lids
Specifications

Specifications

Filtered
Blue
Hinged Rack
Universal Fit
Sterile
Contamination prevention
Low Retention
Aeroject Ultra Filter
Videos
SDS
Documents

Documents

Product Certifications