Learn More
Gibco™ Aeromonas Aminopeptidase Recombinant Protein, PeproTech®
E. coli Recombinant Protein
Brand:  Gibco™ 100-10-1MG
 View more versions of this product
Product Code. 17880783
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
The enzyme (29 KD) aeromonas aminopeptidase is used in the processing of pharmaceutical proteins produced by genetic engineering as well as for physical and structural investigations and for sequence and amino-terminal determinations. This exopeptidase recognizes a specific stop sign at –X- Pro and requires a free a-amino group in the L-configuration. It is therefore suitable for the removal of the redundant N-terminal methionine often added to engineered proteins.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Functional Assay
31.4
2 x 500 μg
-20°C
<1 EU/μg
Unconjugated
MPPITQQATV TAWLPQVDAS QITGTISSLE SFTNRFYTTT SGAQASDWIA SEWQALSASL PNASVKQVSH SGYNQKSVVM TITGSEAPDE WIVIGGHLDS TIGSHTNEQS VAPGADDDASGIAAVTEVIR VLSENNFQPK RSIAFMAYAA EEVGLRGSQD LANQYKSEGK NVVSALQLDM TNYKGSAQDV VFITDYTDSN FTQYLTQLMD EYLPSLTYGF DTCGYACSDH ASWHNAGYPAAMPFESKFND YNPRIHTTQD TLANSDPTGS HAKKFTQLGL AYAIEMGSAT G
≥ 98% by SDS-PAGE gel and HPLC analyses.
protein with no preservative
Aeromonas Aminopeptidase
E. coli
RUO
Bacteria
Recombinant
Lyophilized