missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Aeromonas Aminopeptidase Recombinant Protein, PeproTech®
E. coli Recombinant Protein
£65.00 - £197.00
Specifications
For Use With (Application) | Functional Assay |
---|---|
Formulation | protein with no preservative |
Molecular Weight (g/mol) | 31.4 |
Name | Aeromonas Aminopeptidase |
Source | E. coli |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17850143
|
Gibco™
100-10-100UG |
100 μg |
£65.00
100µg |
Estimated Shipment: 06-11-2024
Log in to see stock. |
|||||
17880783
|
Gibco™
100-10-1MG |
2 x 500 μg | N/A | ||||||
17810793
|
Gibco™
100-10-500UG |
500 μg |
£158.00
500µg |
In Stock
Log in to see stock. |
|||||
Description
The enzyme (29 KD) aeromonas aminopeptidase is used in the processing of pharmaceutical proteins produced by genetic engineering as well as for physical and structural investigations and for sequence and amino-terminal determinations. This exopeptidase recognizes a specific stop sign at –X- Pro and requires a free a-amino group in the L-configuration. It is therefore suitable for the removal of the redundant N-terminal methionine often added to engineered proteins.Specifications
Functional Assay | |
31.4 | |
E. coli | |
RUO | |
Bacteria | |
Recombinant | |
Lyophilized |
protein with no preservative | |
Aeromonas Aminopeptidase | |
-20°C | |
<1 EU/μg | |
Unconjugated | |
MPPITQQATV TAWLPQVDAS QITGTISSLE SFTNRFYTTT SGAQASDWIA SEWQALSASL PNASVKQVSH SGYNQKSVVM TITGSEAPDE WIVIGGHLDS TIGSHTNEQS VAPGADDDASGIAAVTEVIR VLSENNFQPK RSIAFMAYAA EEVGLRGSQD LANQYKSEGK NVVSALQLDM TNYKGSAQDV VFITDYTDSN FTQYLTQLMD EYLPSLTYGF DTCGYACSDH ASWHNAGYPAAMPFESKFND YNPRIHTTQD TLANSDPTGS HAKKFTQLGL AYAIEMGSAT G | |
≥ 98% by SDS-PAGE gel and HPLC analyses. |