missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Aeromonas Aminopeptidase Recombinant Protein, PeproTech®
E. coli Recombinant Protein
Brand: Gibco™ 100-10-01M
This item is not returnable.
View return policy
Description
The enzyme (29 KD) aeromonas aminopeptidase is used in the processing of pharmaceutical proteins produced by genetic engineering as well as for physical and structural investigations and for sequence and amino-terminal determinations. This exopeptidase recognizes a specific stop sign at –X- Pro and requires a free a-amino group in the L-configuration. It is therefore suitable for the removal of the redundant N-terminal methionine often added to engineered proteins.Specifications
Functional Assay | |
31.4 | |
2 x 500 μg | |
-20°C | |
<1 EU/μg | |
Unconjugated | |
MPPITQQATV TAWLPQVDAS QITGTISSLE SFTNRFYTTT SGAQASDWIA SEWQALSASL PNASVKQVSH SGYNQKSVVM TITGSEAPDE WIVIGGHLDS TIGSHTNEQS VAPGADDDASGIAAVTEVIR VLSENNFQPK RSIAFMAYAA EEVGLRGSQD LANQYKSEGK NVVSALQLDM TNYKGSAQDV VFITDYTDSN FTQYLTQLMD EYLPSLTYGF DTCGYACSDH ASWHNAGYPAAMPFESKFND YNPRIHTTQD TLANSDPTGS HAKKFTQLGL AYAIEMGSAT G | |
≥ 98% by SDS-PAGE gel and HPLC analyses. |
Protein with no preservative | |
Aeromonas Aminopeptidase | |
E. coli | |
RUO | |
Bacteria | |
Recombinant | |
Lyophilized |