missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Agarose NA Media
Highly purified agarose for separation of nucleic acids to be used as substrates for enzymes. Cytiva™ Agarose NA Media with low electroendosmosis (EEO) is recommended for most DNA/RNA applications.
Brand: Cytiva 17-0554-03
This item is not returnable.
View return policy
Description
- Gelling temp 34 to 37°C
Specifications
Agarose NA has been prepared to give the optimal combination of these properties: Reliable separation of nucleic acids; low inhibition of enzyme activity on recovered material; and high gel strength. | |
Agarose | |
Agarose with low electroendosmosis (EEO) is recommended for most DNA/RNA applications. Suitable for all analytical separations as well as most preparative methods. | |
60° to 90°C |
White | |
Solid | |
Agarose | |
1 kg |