missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Agarose NA Media
Highly purified agarose for separation of nucleic acids to be used as substrates for enzymes. Cytiva™ Agarose NA Media with low electroendosmosis (EEO) is recommended for most DNA/RNA applications.
£41.06 - £1732.98
Specifications
Color | White |
---|---|
Product Type | Agarose |
Form | Solid |
Material | Agarose |
Melting Temperature | 60° to 90°C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11534925
|
Cytiva
17-0554-02 |
100 g |
£293.00
100g |
Estimated Shipment: 18-07-2024
Log in to see stock. |
|||||
10409764
|
Cytiva
17055401 |
10 g |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
10491455
|
Cytiva
17-0554-03 |
1 kg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Gelling temp 34 to 37°C
Specifications
White | |
Solid | |
60° to 90°C |
Agarose | |
Agarose |