missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Agarose NA Media

Highly purified agarose for separation of nucleic acids to be used as substrates for enzymes. Cytiva™ Agarose NA Media with low electroendosmosis (EEO) is recommended for most DNA/RNA applications.

Brand:  Cytiva 17-0554-02

278.35 GBP valid until 2024-12-31
Use promo code "21631" to get your promotional price.


 View more versions of this product

Product Code. 11534925

  • £322.00 / 100g
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Gelling temp 34 to 37°C
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Agarose NA has been prepared to give the optimal combination of these properties: Reliable separation of nucleic acids; low inhibition of enzyme activity on recovered material; and high gel strength.
Agarose
Agarose with low electroendosmosis (EEO) is recommended for most DNA/RNA applications. Suitable for all analytical separations as well as most preparative methods.
60° to 90°C
White
Solid
Agarose
100 g
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.