Learn More
Brady™ Aggressive Adhesive Die-cut Multipurpose Nylon Label Tape for M6/M7 Printers
Aggressive adhesive and high pliability are the hallmarks of this nylon cloth label material designed to meet a broad array of identification requirements, from control panels to liquid nitrogen environments.
Brand:  Brady™ 173395
Additional Details : Weight : 0.08900kg
 View more versions of this product
Product Code. 17818844
  • £48.20 / Pack of 250
Estimated Shipment: 08-08-2024
to see stock.
Add to basket
Description
Description
  • Highly pliable material conforms tightly to curved surfaces such as wires and cables while also adhering to stainless steel, textured ABS, polypropylene and many other material types
  • Resists exposure to cold temperatures, chemicals, greases, oils, cleaners, and detergents
  • Low profile label material adheres to smooth, rough, or powder coated surfaces or even materials with low surface energy
  • UL Recognized to UL969, CSA to C222 No 015-95, RoHS compliant to 2011/65/EU (Pass 2015/863)
  • Made of B-499 nylon cloth material
Specifications
Specifications
Autoclavable
Permanent Acrylic
White
Die-cut
Single-side Printable
Labels
Component and Equipment Labeling, General Identification, Laboratory Labeling
No
Square
-40°C to +90°C
0.166 mm
1 in.
25.4 mm
Halogen-free (DIN VDE 0472 Part 815)
None
Matte
Nylon
Thermal Transfer
Non-sterile
M710, BMP71
250 Labels
-40°F to +194°F
0.0065 in.
1 x 1 in.
1 in.
25.4 mm