Learn More
Brady™ Aggressive Adhesive Die-cut Multipurpose Nylon Label Tape for M6/M7 Printers
Aggressive adhesive and high pliability are the hallmarks of this nylon cloth label material designed to meet a broad array of identification requirements, from control panels to liquid nitrogen environments.
£33.25 - £101.00
Specifications
Autoclavable | Autoclavable |
---|---|
Certifications/Compliance | Halogen-free (DIN VDE 0472 Part 815) |
Adhesive Type | Permanent Acrylic |
Coating | None |
Color | White |
Product Code | Brand | Dimensions (LxW) | For Use With (Equipment) | Height (Metric) | Shape | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (LxW) | For Use With (Equipment) | Height (Metric) | Shape | Price | Quantity & Availability | ||
17858124
|
Brady™
173311 |
0.5 x 0.5 in. | M710, BMP71 | 12.7 mm | Square |
£59.05
Pack of 500 |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||
17838454
|
Brady™
174349 |
0.5 x 0.5 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | 12.7 mm | Square |
£63.15
Pack of 500 |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||
17818844
|
Brady™
173395 |
1 x 1 in. | M710, BMP71 | 25.4 mm | Square |
£48.20
Pack of 250 |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||
17828404
|
Brady™
174225 |
1 x 1 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | 25.4 mm | Square |
£51.50
Pack of 250 |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||
17868974
|
Brady™
174594 |
0.25 x 1.5 in. | M710, BMP71 | 6.35 mm | Rectangle |
£62.70
Pack of 750 |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||
17878194
|
Brady™
173307 |
0.25 x 1.9 in. | M710, BMP71 | 6.35 mm | Rectangle |
£72.35
Pack of 750 |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||
17811389
|
Brady™
173790 |
0.25 x 1.5 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | 6.35 mm | Rectangle |
£59.44
Pack of 750 |
Estimated Shipment: 09-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
17888284
|
Brady™
174357 |
0.25 x 1.9 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | 6.35 mm | Rectangle |
£76.70
Pack of 750 |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||
17838864
|
Brady™
174650 |
0.5 x 0.9 in. | M710, BMP71 | 12.7 mm | Rectangle |
£93.80
Pack of 500 |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||
17818384
|
Brady™
174370 |
0.5 x 0.9 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | 12.7 mm | Rectangle |
£101.00
Pack of 500 |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||
17888294
|
Brady™
174184 |
0.75 x 0.5 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | 19.05 mm | Rectangle |
£57.65
Pack of 500 |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||
17818464
|
Brady™
174218 |
0.75 x 1 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | 19.05 mm | Rectangle |
£47.60
Pack of 250 |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||
17801499
|
Brady™
174273 |
0.75 x 1.5 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | 19.05 mm | Rectangle |
£59.74
Pack of 250 |
Estimated Shipment: 09-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
17868194
|
Brady™
174201 |
1 x 0.375 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | 25.4 mm | Rectangle |
£68.90
Pack of 500 |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||
17879024
|
Brady™
173415 |
1 x 0.375 in. | M710, BMP71 | 25.4 mm | Rectangle |
£64.45
Pack of 500 |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||
17878784
|
Brady™
173401 |
1 x 0.75 in. | M710, BMP71 | 25.4 mm | Rectangle |
£44.50
Pack of 250 |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||
17898364
|
Brady™
174103 |
1 x 0.5 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | 25.4 mm | Rectangle |
£44.70
Pack of 500 |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||
17888114
|
Brady™
174280 |
1 x 1.5 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | 25.4 mm | Rectangle |
£65.60
Pack of 250 |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||
17898254
|
Brady™
174354 |
1 x 1.9 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | 25.4 mm | Rectangle |
£98.30
Pack of 250 |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||
17888434
|
Brady™
173310 |
1 x 1.9 in. | M710, BMP71 | 25.4 mm | Rectangle |
£92.70
Pack of 250 |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||
17818924
|
Brady™
173355 |
1.5 x 0.5 in. | M710, BMP71 | 38.1 mm | Rectangle |
£60.05
Pack of 500 |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||
17828844
|
Brady™
174265 |
1.5 x 0.5 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | 38.1 mm | Rectangle |
£64.20
Pack of 500 |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||
17888904
|
Brady™
173347 |
1.5 x 0.75 in. | M710, BMP71 | 38.1 mm | Rectangle |
£62.00
Pack of 250 |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||
17898924
|
Brady™
173340 |
1.5 x 1 in. | M710, BMP71 | 38.1 mm | Rectangle |
£61.35
Pack of 250 |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||
17831379
|
Brady™
173769 |
1 x 4 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | 25.4 mm | Rectangle |
£43.71
Pack of 100 |
Estimated Shipment: 09-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
17868344
|
Brady™
174086 |
1.25 x 1.5 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | 31.75 mm | Rectangle |
£83.85
Pack of 250 |
Estimated Shipment: 06-09-2024
Log in to see stock. |
||
17899034
|
Brady™
174575 |
2.5 x 1 in. | M710, BMP71 | 63.5 mm | Rectangle |
£35.90
Pack of 100 |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||
17891569
|
Brady™
173756 |
2.5 x 1 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | 63.5 mm | Rectangle |
£34.00
Pack of 100 |
Estimated Shipment: 09-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
17888464
|
Brady™
174085 |
0.375 x 0.825 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | 9.53 mm | Rectangle |
£33.25
Pack of 500 |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||
17888734
|
Brady™
173427 |
0.75 x 0.5 in. | M710, BMP71 | 19.05 mm | Rectangle |
£53.90
Pack of 500 |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||
Description
- Highly pliable material conforms tightly to curved surfaces such as wires and cables while also adhering to stainless steel, textured ABS, polypropylene and many other material types
- Resists exposure to cold temperatures, chemicals, greases, oils, cleaners, and detergents
- Low profile label material adheres to smooth, rough, or powder coated surfaces or even materials with low surface energy
- UL Recognized to UL969, CSA to C222 No 015-95, RoHS compliant to 2011/65/EU (Pass 2015/863)
- Made of B-499 nylon cloth material
Specifications
Autoclavable | |
Permanent Acrylic | |
White | |
Die-cut | |
Single-side Printable | |
Labels | |
Component and Equipment Labeling, General Identification, Laboratory Labeling | |
-40°F to +194°F | |
0.0065 in. |
Halogen-free (DIN VDE 0472 Part 815) | |
None | |
Matte | |
Nylon | |
Thermal Transfer | |
Non-sterile | |
No | |
-40°C to +90°C | |
0.166 mm |