Learn More
Brady™ Aggressive Adhesive Multipurpose Polyester Labels
Easily label components with the thermal transfer, glossy permanent polyester label material (B-422). Print almost anywhere with a durable Brady™ handheld label printer.
Brand:  Brady™ 170732
 View more versions of this product
Product Code. 17990680
  • £43.85 / Each
Explore more special offers
Description
Description
  • Designed for a wide range of applications such as labeling equipment, circuit boards, panel components and barcodes
  • Constructed to withstand abrasions, extreme temperatures and solvents such as alcohol, oil and water
  • Made with a permanent acrylic adhesive that sticks to rough, smooth and powder-coated flat surfaces
Specifications
Specifications
UL Recognized to UL969, CSA to C22.2 No.0.15-95, RoHS Compliant to 2011/65/EU (Pass 2015/863)
Permanent Acrylic
Gloss
Polyester
Non-sterile
BMP41, BMP51, M511
-40°F to +212°F
25 ft. x 0.5 in.
0.5 in.
M4C-500-422
Black on White
Continuous
Labels
Asset Tracking, Barcode Labeling, Component and Equipment Labeling, Data and Telecommunications Labeling, Electrical Labeling, General Identification, Inventory and Inspection Labeling, Wire and Cable Labeling
1 Tape/Cartridge
0.004 in.
25 ft.
1.3 cm