missing translation for 'onlineSavingsMsg'
Learn More

Trajan™ Split / Splitless Tapered FocusLiner for Agilent

The unique deactivation process of SGE inlet liners provides excellent analysis for a wide range of compounds

Brand:  Trajan™ 09200301

Code : 13

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 12917141

  • £38.20 / Each
Estimated Shipment: 16-08-2024
to see stock.
Add to basket

Description

Description

  • Each batch of liners tested for activity
  • Manufactured from borosilicate glass
  • Color coded by geometry for easy identification
  • For use with Agilent instruments
Specifications

Specifications

GC Inlet Liner, Split/Splitless Tapered FocusLiner
78.5 mm
6.3 mm
1 Ea.
4 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers