Learn More
Tocris Bioscience™ AGN 193109
High affinity pan-RAR antagonist
Brand: Tocris Bioscience™ 5758/10
226.34 GBP valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Description
AGN 193109 is a high affinity pan-retinoic acid receptor (RAR) antagonist (Kd values are 2, 2 and 3 nM at RARα, RARβ and RARγ, respectively). Exhibits no significant affinity for retinoic X receptors. AGN 193109 promotes the differentiation from mouse embryonic stem cells into paraxial mesoderm. AGN 193109 suppresses primary human cardiovascular smooth muscle cell calcification.AGN 193109 synthesized to Ancillary Material Grade also available.
Specifications
Antagonist | |
171746-21-7 | |
392.49g/mol | |
Retinoic Acid Receptor Antagonists |
Retinoic Acid Receptor | |
C28H24O2 | |
Store at -20°C | |
10 mg |
For Research Use Only