Learn More
Abeba™ Air-Cushion Sole 2600 Shoes
Antistatic with air cushion PU-sole and high slip resistance. Sanitized™ treated.
Brand: Abeba™ 2600/46
Additional Details : Weight : 0.65000kg
Description
- CE EN ISO 20347:2012 O2 SRA
- Grained leather
- White
- Slip resistance PU outsole sole
- Replaceable insole (article no. 3551)
- Sanitized treated
- Antistatic
- Lining microfiber, respiration absorbing
- Instep area with lacing
Specifications
CE EN ISO 20347:2012 O2 SRA | |
Microfiber | |
46 | |
Polyurethane |
White | |
Leather | |
Slip-resistant | |
2600 |