Learn More
Asecos™ Air Purifier PURIFIAIR.620
Air Purifier PURIFIAIR.620 reduces indoor viral loads and pollutant levels with a multi-component filter system.
Brand:  Asecos™ 39326
 View more versions of this product
Product Code. 17222534
  • POA / Each
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Explore more special offers
Description
Description
  • Asecos PURIFIAIR.620 can significantly reduce viral loads and pollution in indoor areas
    • Removes mould spores, pollen, and allergens
    • Eliminates germs and bacteria and neutralises them
    • Continuously filters out fine dust particles
    • Removes unpleasant odours and harmful gases from indoor atmospheres
    • Built-in PM1.0 sensor and colour display allow users to quickly identify and read off the indoor atmosphere particle concentration in four levels from green (low) to red (high) concentration
    • Built-in combination of pre-filters, an E12 EPA Filter and H14 HEPA Filter in accordance with EN 1822 has an efficiency of up to 99.995% while keeping out particles from 0.1 bis 0.3 μm in size
    • H14 HEPA filters are used in areas where protection against infection plays a key role
    • The mobile design allows the purifier to be used flexibly in different areas
  • 1 device handles a volume flow rate of 160 bis to 620 m³/h (5 operating levels) and is suitable for approx. 100 m³ room volume (corresponding to a surface area of 40 m² and a room height of 2.5 m)
    • A larger number of devices should be used in larger rooms
    • Fresh air must be supplied to the rooms via natural or technical ventilation
    • Air purifiers should not be considered a substitute for a fresh air supply
    • Efficient results over time only if the filter is replaced at regularly according to the electronic display
    • Large filter units ensure a long service life
    • Low energy requirements result in low usage costs
Specifications
Specifications
Prefilter, Cold Catalysis Filter, Activated Carbon Filter, High-performance Particle Filter
2.867 Kg
509 mm
Replacement Filter
AIRPURIFIAIR.620
162 mm
402 mm
EN 1822
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Asecos™ Air Purifier PURIFIAIR.620 > Asecos Air Purifier PURIFIAIR.620; Product Type: Replacement Filter; For Use With (Equipment): PURIFIAIR.620