Learn More
Syngene™ Airclean™ Process Protection Microscope Hood
GREENER_CHOICE
Process Protection with UVTect™ Microprocessor and Microscope Sash
Brand:  Syngene™ SYN-48PPUVC-E
8307.08 GBP valid until 2024-12-31
Use promo code "21658" to get your promotional price.
 View more versions of this product
Product Code. 16552972
  • £9400.00 / Each
Explore more special offers
Description
Description
  • AirClean™ Systems AC600 Series Vertical Laminar Flow Workstation
  • Laminar air flow is described as the smooth, fluid, parallel flow of air in a consistent direction
  • Ideal for applications that are not hazardous to the operator but require a clean workspace
  • A compact, light-weight design allows for multiple clean zones within a laboratory
  • Two-stage filtration provides clean, Class 100 laminar flow air for process protection
  • Room air first passes through an electrostatically charged pre-filter followed by a HEPA filter resulting in ISO 5 clean air
  • Independently certified to meet or exceed applicable clean bench standards
Specifications
Specifications
Laminar Flow
81.28 cm
121.92 cm
61 cm
48 in.