Learn More
Invitrogen™ ProcartaPlex™ Human Akt Pathway Panel, total, 8plex
The ProcartaPlex Human Akt Pathway Panel, total, 8plex, enables simultaneous quantitation of the total levels from eight key proteins included in the Akt cell signaling pathway.
Brand:  Invitrogen™ EPX080-97000-901
Product Code. 18456698
  • £2115.00 / Pack of 96
Explore more special offers
Description
Show More
Specifications
Specifications
Multiplex Panel
Human
Magnetic
See Certificate of Analysis
Luminex™ Instruments
Akt, PKB, Protein kinase B
Akt
ProcartaPlex™
Cell lysates (prediluted): 25 μL
Wet Ice
96 Tests
Non-Combinable
See Certificate of Analysis
Multiplex Kit
Akt
207
8-plex
Cell lysates
P31749
Signaling
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number430812-003 Certificate TypeCertificate of Analysis Date24/04/2025 Catalog Number
Lot Number430812-002 Certificate TypeCertificate of Analysis Date10/04/2025 Catalog Number
Lot Number430812-001 Certificate TypeCertificate of Analysis Date12/11/2024 Catalog NumberEPX080-97000-901
Lot Number404605-003 Certificate TypeCertificate of Analysis Date02/07/2024 Catalog NumberEPX080-97000-901
Lot Number404605-002 Certificate TypeCertificate of Analysis Date05/06/2024 Catalog NumberEPX080-97000-901
Special Offers
Special Offers
Product Content Correction
Product Title
Invitrogen™ ProcartaPlex™ Human Akt Pathway Panel, total, 8plex > 96 Tests