missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AKT Rabbit anti-Human, Mouse, Rat, Zebrafish, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10176-2-AP-20UL
This item is not returnable.
View return policy
Description
The serine-threonine protein kinase encoded by the AKT1 gene is catalytically inactive in serum-starved primary and immortalized fibroblasts. AKT1 and the related AKT2 are activated by platelet-derived growth factor. The activation is rapid and specific, and it is abrogated by mutations in the pleckstrin homology domain of AKT1. It was shown that the activation occurs through phosphatidylinositol 3-kinase. In the developing nervous system AKT is a critical mediator of growth factor-induced neuronal survival. Survival factors can suppress apoptosis in a transcription-independent manner by activating the serine/threonine kinase AKT1, which then phosphorylates and inactivates components of the apoptotic machinery. Mutations in this gene have been associated with the Proteus syndrome. Multiple alternatively spliced transcript variants have been found for this gene.Specifications
AKT | |
Polyclonal | |
Unconjugated | |
AKT1 | |
AKT, AKT1, AKT2, AKT3, PKB, PKB ALPHA, PRKBA, Protein kinase B, Proto oncogene c Akt, RAC, RAC ALPHA, RAC PK alpha | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10000, 101910198, 11651, 11652, 207, 208, 23797, 24185, 25233, 29414, 378972, 560549 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Flow Cytometry, Immunofluorescence, Immunocytochemistry | |
0.4 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P31749, P31750, P31751, P47196, P47197, Q60823, Q63484, Q9WUA6, Q9Y243 | |
akt1, akt2, Akt3, akt3a | |
AKT Fusion Protein Ag0213 | |
20 μL | |
Primary | |
Rat, Mouse, Human, Zebrafish | |
Antibody | |
IgG |