missing translation for 'onlineSavingsMsg'
Learn More

AKT Rabbit anti-Human, Mouse, Rat, Zebrafish, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

£135.00 - £349.00

Specifications

Antigen AKT
Concentration 0.4 mg/mL
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Flow Cytometry, Immunofluorescence, Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16866383
View Documents Promotion Details
Proteintech
10176-2-AP-20UL
20 μL
£135.00
20µL
Please to purchase this item. Need a web account? Register with us today!
16856383
View Documents Promotion Details
Proteintech
10176-2-AP-150UL
150 μL
£349.00
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The serine-threonine protein kinase encoded by the AKT1 gene is catalytically inactive in serum-starved primary and immortalized fibroblasts. AKT1 and the related AKT2 are activated by platelet-derived growth factor. The activation is rapid and specific, and it is abrogated by mutations in the pleckstrin homology domain of AKT1. It was shown that the activation occurs through phosphatidylinositol 3-kinase. In the developing nervous system AKT is a critical mediator of growth factor-induced neuronal survival. Survival factors can suppress apoptosis in a transcription-independent manner by activating the serine/threonine kinase AKT1, which then phosphorylates and inactivates components of the apoptotic machinery. Mutations in this gene have been associated with the Proteus syndrome. Multiple alternatively spliced transcript variants have been found for this gene.
Specifications

Specifications

AKT
Immunohistochemistry (Paraffin), Immunoprecipitation, Flow Cytometry, Immunofluorescence, Immunocytochemistry
Unconjugated
Rabbit
Rat, Mouse, Human, Zebrafish
P31749, P31750, P31751, P47196, P47197, Q60823, Q63484, Q9WUA6, Q9Y243
10000, 101910198, 11651, 11652, 207, 208, 23797, 24185, 25233, 29414, 378972, 560549
AKT Fusion Protein Ag0213
Primary
-20°C
AKT1
0.4 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
AKT, AKT1, AKT2, AKT3, PKB, PKB ALPHA, PRKBA, Protein kinase B, Proto oncogene c Akt, RAC, RAC ALPHA, RAC PK alpha
akt1, akt2, Akt3, akt3a
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications