missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Alcian Blue 8GX, certified
£166.83 - £782.90
Chemical Identifiers
CAS | 33864-99-2 |
---|---|
Molecular Formula | C56H68Cl4CuN16S4 |
Molecular Weight (g/mol) | 1298.86 |
MDL Number | MFCD00010720 |
InChI Key | XRWQTDIELZDCPS-UHFFFAOYSA-J |
Synonym | Ingrain Blue 1 |
PubChem CID | 17749099 |
IUPAC Name | (9Z,18Z,26Z,35Z)-4,14,22,32-tetrakis({[(dimethylamino)(dimethyliminiumyl)methyl]sulfanyl}methyl)-9,18,27,36,37,39,40,41-octaaza-38-cupradecacyclo[17.17.3.1¹⁰,¹⁷.1²⁸,³⁵.0²,⁷.0⁸,³⁷.0¹¹,¹⁶.0²⁰,²⁵.0²⁶,³⁹.0²⁹,³⁴]hentetraconta-1,3,5,7,9,11(16),12,14,17(41),18,20,22,24,26,28(40),29(34),30,32,35-nonadecaene-38,38-bis(ylium) tetrachloride |
SMILES | [Cl-].[Cl-].[Cl-].[Cl-].CN(C)C(SCC1=CC2=C(C=C1)\C1=N\C3=C4C=CC(CSC(N(C)C)=[N+](C)C)=CC4=C4\N=C5/N=C(/N=C6\N([Cu++]N34)\C(=N/C2=N1)C1=CC(CSC(N(C)C)=[N+](C)C)=CC=C61)C1=C5C=C(CSC(N(C)C)=[N+](C)C)C=C1)=[N+](C)C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10786953
|
Thermo Scientific Chemicals
400460100 |
10 g | Glass bottle |
£166.83
10g |
In Stock
Log in to see stock. |
||||
10577162
|
Thermo Scientific Chemicals
400460250 |
25 g | Glass bottle |
£297.48
25g |
In Stock
Log in to see stock. |
||||
10619263
|
Thermo Scientific Chemicals
400461000 |
100 g | Glass bottle |
£782.90
100g |
In Stock
Log in to see stock. |
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
33864-99-2 | |
1298.86 | |
XRWQTDIELZDCPS-UHFFFAOYSA-J | |
17749099 | |
[Cl-].[Cl-].[Cl-].[Cl-].CN(C)C(SCC1=CC2=C(C=C1)\C1=N\C3=C4C=CC(CSC(N(C)C)=[N+](C)C)=CC4=C4\N=C5/N=C(/N=C6\N([Cu++]N34)\C(=N/C2=N1)C1=CC(CSC(N(C)C)=[N+](C)C)=CC=C61)C1=C5C=C(CSC(N(C)C)=[N+](C)C)C=C1)=[N+](C)C |
C56H68Cl4CuN16S4 | |
MFCD00010720 | |
Ingrain Blue 1 | |
(9Z,18Z,26Z,35Z)-4,14,22,32-tetrakis({[(dimethylamino)(dimethyliminiumyl)methyl]sulfanyl}methyl)-9,18,27,36,37,39,40,41-octaaza-38-cupradecacyclo[17.17.3.1¹⁰,¹⁷.1²⁸,³⁵.0²,⁷.0⁸,³⁷.0¹¹,¹⁶.0²⁰,²⁵.0²⁶,³⁹.0²⁹,³⁴]hentetraconta-1,3,5,7,9,11(16),12,14,17(41),18,20,22,24,26,28(40),29(34),30,32,35-nonadecaene-38,38-bis(ylium) tetrachloride |
Specifications
148.0°C | |
50% min. | |
MFCD00010720 | |
Ingrain Blue 1 | |
XRWQTDIELZDCPS-UHFFFAOYSA-J | |
[Cl-].[Cl-].[Cl-].[Cl-].CN(C)C(SCC1=CC2=C(C=C1)\C1=N\C3=C4C=CC(CSC(N(C)C)=[N+](C)C)=CC4=C4\N=C5/N=C(/N=C6\N([Cu++]N34)\C(=N/C2=N1)C1=CC(CSC(N(C)C)=[N+](C)C)=CC=C61)C1=C5C=C(CSC(N(C)C)=[N+](C)C)C=C1)=[N+](C)C | |
1298.86 | |
1298.88 | |
Crystalline Powder |
33864-99-2 | |
C56H68Cl4CuN16S4 | |
15, 212 | |
Solubility in water: moderately soluble. Other solubilities: soluble in ethanol, cellosolve, ethylene glycol, practically insoluble in xylene | |
Authentic | |
(9Z,18Z,26Z,35Z)-4,14,22,32-tetrakis({[(dimethylamino)(dimethyliminiumyl)methyl]sulfanyl}methyl)-9,18,27,36,37,39,40,41-octaaza-38-cupradecacyclo[17.17.3.1¹⁰,¹⁷.1²⁸,³⁵.0²,⁷.0⁸,³⁷.0¹¹,¹⁶.0²⁰,²⁵.0²⁶,³⁹.0²⁹,³⁴]hentetraconta-1,3,5,7,9,11(16),12,14,17(41),18,20,22,24,26,28(40),29(34),30,32,35-nonadecaene-38,38-bis(ylium) tetrachloride | |
17749099 | |
Purple | |
Alcian Blue 8GX |
RUO – Research Use Only