missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ALDH1A3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25167-1-AP-150UL
This item is not returnable.
View return policy
Description
ALDH1A3 are aldehyde dehydrogenase isozymes which are thought to play a major role in the detoxification of aldehydes generated by alcohol metabolism and lipid peroxidation. The enzyme encoded by the ALDH1A3 gene uses retinal as a substrate, either in a free or cellular retinol-binding protein form.Specifications
ALDH1A3 | |
Polyclonal | |
Unconjugated | |
ALDH1A3 | |
Aldehyde dehydrogenase 6, ALDH1A3, ALDH1A6, ALDH6, RALDH 3, RALDH3, Retinaldehyde dehydrogenase 3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
220, 56847 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.39 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P47895, Q9JHW9 | |
Aldh1a3 | |
ALDH1A3 Fusion Protein Ag18605 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction