missing translation for 'onlineSavingsMsg'
Learn More

ALDH1A3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

£135.00 - £349.00

Specifications

Antigen ALDH1A3
Concentration 0.39 mg/mL
Applications Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16843725
View Documents
Proteintech
25167-1-AP-20UL
20 μL
£135.00
20µL
Please to purchase this item. Need a web account? Register with us today!
16833725
View Documents
Proteintech
25167-1-AP-150UL
150 μL
£349.00
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

ALDH1A3 are aldehyde dehydrogenase isozymes which are thought to play a major role in the detoxification of aldehydes generated by alcohol metabolism and lipid peroxidation. The enzyme encoded by the ALDH1A3 gene uses retinal as a substrate, either in a free or cellular retinol-binding protein form.
Specifications

Specifications

ALDH1A3
Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence
Unconjugated
Rabbit
Mouse, Human
P47895, Q9JHW9
220, 56847
ALDH1A3 Fusion Protein Ag18605
Primary
-20°C
ALDH1A3
0.39 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Aldehyde dehydrogenase 6, ALDH1A3, ALDH1A6, ALDH6, RALDH 3, RALDH3, Retinaldehyde dehydrogenase 3
Aldh1a3
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
ALDH1A3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.