Learn More
ALDH1A3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 25167-1-AP-20UL
 View more versions of this product
Product Code. 16843725
  • £135.00 / 20µL
Explore more special offers
Description
Description
ALDH1A3 are aldehyde dehydrogenase isozymes which are thought to play a major role in the detoxification of aldehydes generated by alcohol metabolism and lipid peroxidation. The enzyme encoded by the ALDH1A3 gene uses retinal as a substrate, either in a free or cellular retinol-binding protein form.
Specifications
Specifications
ALDH1A3
Polyclonal
Unconjugated
ALDH1A3
Aldehyde dehydrogenase 6, ALDH1A3, ALDH1A6, ALDH6, RALDH 3, RALDH3, Retinaldehyde dehydrogenase 3
Rabbit
Antigen Affinity Chromatography
RUO
220, 56847
-20°C
Liquid
Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence
0.39 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P47895, Q9JHW9
Aldh1a3
ALDH1A3 Fusion Protein Ag18605
20 μL
Primary
Mouse, Human