Learn More
Molecular Probes™ Alexa Fluor™ 488 Azide (Alexa Fluor™ 488 5-Carboxamido-(6-Azidohexanyl), Bis(Triethylammonium Salt)), 5-isomer
The green-fluorescent Alexa Fluor™ 488 azide is reactive with terminal alkynes via a copper-catalyzed click reaction.
Brand: Molecular Probes™ A10266
279.68 GBP valid until 2025-04-18
Use promo code "24107" to get your promotional price.
Description
The green-fluorescent Alexa Fluor™ 488 azide is reactive with terminal alkynes via a copper-catalyzed click reaction. In addition to being an exceedingly bright and photostable fluorophore for use with flow cytometry, microscopy and HCS, Alexa Fluor™ 488 can also be utilized as a bio-orthogonal or biologically unique hapten for use in applications requiring signal amplification.
Stock concentrations of 1 to 10 mM in high-quality anhydrous DMSO are recommended.

Specifications
Solid | |
Azide | |
DMF (Dimethylformamide), DMSO (Dimethylsulfoxide) | |
Alkyne | |
Room Temperature | |
Amine, Azide | |
Alexa Fluor | |
Alexa Fluor™, Molecular Probes™ | |
495 nm |
0.5 mg | |
861.04 g/mol | |
Store at ≤-20°C, desiccate, and protect from light. | |
Fluorescence | |
Click-iT™ | |
Alexa Fluor™ 488 | |
Green | |
519 nm |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Special Offers
-
Multiple Manufacturers
Powering Proteomics
Advance your proteomics research with essential toolsPromo Code: 25977Expires: 2025-05-31
New Offer Save Up to 92%
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.