missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ Alexa Fluor™ Antibody Labeling Kits

Multiplex with brighter, photostable, Alexa Fluor Antibody Labeling Kits without the intramolecular quenching. Conjugate antibodies to 9 available fluorophores for flow cytometry, ICC, IHC, IF, ELISA or HCS. Each kit contains enough dye to conjugate 100 μg per reaction with 5 reactions per kit.

Brand:  Molecular Probes™ A20185

427.12 GBP valid until 2025-03-28
Use promo code "24107" to get your promotional price.


517.00 GBP valid until 2025-05-31
Use promo code "25977" to get your promotional price.


 View more versions of this product

Product Code. 10693833

  • £582.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

For Research Use Only. All usage must comply with product instructions.
 
This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

Labeling Kit
Alexa Fluor™ 594 Antibody Labeling Kit comes with 5 vials of reactive dye, sodium bicarbonate, purification resin, spin columns, and collections tubes.

Store at 2°C to 8°C, protected from light. Component A may be stored from -20°C to +8°C.
590/616 nm
100 μg
Alexa Fluor™
DMSO (Dimethylsulfoxide)
Red
Alexa Fluor
1 kit
Fluorescence
Conjugation-based
Room Temperature
Amine
Alexa Fluor 594
Antibodies, Proteins
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2780656 Certificate TypeCertificate of Analysis Date03/01/2024 Catalog NumberA20185
Lot Number2622315 Certificate TypeCertificate of Analysis Date04/05/2023 Catalog NumberA20185
Lot Number2480136 Certificate TypeCertificate of Analysis Date27/06/2022 Catalog NumberA20185
Lot Number2381704 Certificate TypeCertificate of Analysis Date23/09/2021 Catalog NumberA20185
Lot Number2335863 Certificate TypeCertificate of Analysis Date29/04/2021 Catalog NumberA20185
Special Offers

Special Offers

  • Multiple Manufacturers
    Advance your proteomics research with essential tools
    Promo Code: 25977

    Expires2025-05-31

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Molecular Probes™ Alexa Fluor™ Antibody Labeling Kits > Alexa Fluor™ 594

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.