Learn More
S Murray™ Samco™ All Glass Luer-Tip Syringe
Use these syringes for a variety of research applications. S Murray™ All Glass Luer-Tip Syringes are graduated and feature glass luer fittings.
Brand: S Murray™ G811/02
Description
Use these syringes for a variety of research applications. S Murray™ All Glass Luer-Tip Syringes are graduated and feature glass luer fittings.
- Syringe with glass luer fitting
- Fully interchangeable
Specifications
CE | |
All Glass Luer-tip Syringe | |
2 mL | |
Yes |
Glass | |
1/Ea. | |
Luer Tip |