missing translation for 'onlineSavingsMsg'
Learn More

Brady™ All-weather Vinyl Labels

Make labels that withstand up to 10 years of outdoor abuse with this low-shrink vinyl label material (B-595). Print almost anywhere with a durable Brady handheld label maker.

Brand:  Brady™ 176803

Additional Details : Weight : 0.12000kg

 View more versions of this product

Product Code. 17871739

  • £41.18 / Each
Estimated Shipment: 30-09-2024
to see stock.

Description

Description

  • Make labels that withstand up to 10 years of outdoor abuse with this low-shrink vinyl label material (B-595). Print almost anywhere with a durable Brady handheld label maker.
  • Versatile: Ideal for labeling pipes, equipment, shelves and other industrial surfaces
  • Tough: Designed to withstand prolonged exposure to UV light, chemicals, greases and oils, salt spray, cleaners and detergents, and grime
  • Adhesive: Made with high-tack, permanent acrylic adhesive for use on rough, highly-textured, dirty and smooth surfaces alike, especially in cold temperatures
  • Long-lasting: Weather-resistant labels designed to last up to 10 years in an outdoor environment
Specifications

Specifications

Permanent Acrylic
Black on White
Label
Front
Labels
Component and Equipment Labelling, Electrical Labelling, Facility Identification, Laboratory Labelling, Lean and 5S Labelling, Lockout Tagout, Security Identification, Pipe and Valve Marking
1
1 Ea.
-40°C to +82°C
12.70 x 12.70 mm
12.70 mm
12.70 mm
None
Gloss
Vinyl
Thermal Transfer
Non-sterile
BMP41, BMP51, M511 Printers
280 Labels/Box
-40°F to +180°F
0.102 mm
0.5 in.
0.5 in.
12.70 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers