Learn More
Allantoin, 98%, Thermo Scientific Chemicals
£51.50 - £726.00
Chemical Identifiers
CAS | 97-59-6 |
---|---|
Molecular Formula | C4H6N4O3 |
Molecular Weight (g/mol) | 158.117 |
MDL Number | MFCD00005260 |
InChI Key | POJWUDADGALRAB-UHFFFAOYSA-N |
Synonym | allantoin, 5-ureidohydantoin, glyoxyldiureide, 1-2,5-dioxoimidazolidin-4-yl urea, glyoxyldiureid, allantol, cordianine, sebical, alantan, avc/dienestrolcream |
PubChem CID | 204 |
ChEBI | CHEBI:15676 |
IUPAC Name | (2,5-dioxoimidazolidin-4-yl)urea |
SMILES | C1(C(=O)NC(=O)N1)NC(=O)N |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11403047
|
Thermo Scientific Alfa Aesar
A15571.30 |
250 g |
£51.50
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11493037
|
Thermo Scientific Alfa Aesar
A15571.0B |
1000 g |
£172.00
1000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11413047
|
Thermo Scientific Alfa Aesar
A15571.0I |
5000 g |
£726.00
5000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Allantoin is a useful biomarker for oxidative stress. It is a reactant for Synthesis of sulofenur analogs for use as antitumor agents. It is used as a cosmetic agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsAllantoin is a useful biomarker for oxidative stress. It is a reactant for Synthesis of sulofenur analogs for use as antitumor agents. It is used as a cosmetic agent.
Solubility
Slightly soluble in water. Freely soluble in alkalis
Notes
Store in a cool, dry conditions in a well sealed container. Incompatible with oxidizing agent.
Chemical Identifiers
97-59-6 | |
158.117 | |
POJWUDADGALRAB-UHFFFAOYSA-N | |
204 | |
(2,5-dioxoimidazolidin-4-yl)urea |
C4H6N4O3 | |
MFCD00005260 | |
allantoin, 5-ureidohydantoin, glyoxyldiureide, 1-2,5-dioxoimidazolidin-4-yl urea, glyoxyldiureid, allantol, cordianine, sebical, alantan, avc/dienestrolcream | |
CHEBI:15676 | |
C1(C(=O)NC(=O)N1)NC(=O)N |
Specifications
97-59-6 | |
C4H6N4O3 | |
102364 | |
allantoin, 5-ureidohydantoin, glyoxyldiureide, 1-2,5-dioxoimidazolidin-4-yl urea, glyoxyldiureid, allantol, cordianine, sebical, alantan, avc/dienestrolcream | |
POJWUDADGALRAB-UHFFFAOYSA-N | |
(2,5-dioxoimidazolidin-4-yl)urea | |
204 | |
158.12 | |
Allantoin |
∼230°C (decomposition) | |
MFCD00005260 | |
14,258 | |
Slightly soluble in water. Freely soluble in alkalis | |
C1(C(=O)NC(=O)N1)NC(=O)N | |
158.117 | |
CHEBI:15676 | |
98% |
Safety and Handling
missing translation for 'einecsNumber' : 202-592-8
missing translation for 'rtecsNumber' : YT1600000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only