Learn More
Allihn Condenser with Ground Cone and Socket
Ideal for reflux applications.
Brand: Quickfit CX7/33/SC
195.90 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- Inner bulb sections provide increased cooling surface
- With detachable plastic screwthread connectors for easy and safe attachment of flexible tubing
Specifications
Ideal for reflux work | |
545 mm | |
Borosilicate Glass |
545 mm | |
24/29 | |
Allihn Condenser |