Learn More
Allyl mercaptan, tech., 70%, remainder mainly sulfide, Thermo Scientific Chemicals
£87.70
Chemical Identifiers
CAS | 870-23-5 |
---|---|
Molecular Formula | C3H6S |
Molecular Weight (g/mol) | 74.14 |
MDL Number | MFCD00004894 |
InChI Key | ULIKDJVNUXNQHS-UHFFFAOYSA-N |
Synonym | allyl mercaptan, 2-propene-1-thiol, allylthiol, allylmercaptan, allyl sulfhydrate, unii-1x587iby09, 2-propenyl mercaptan, 2-propenyl-1-thiol, ch2=chch2sh, allyl thiol |
PubChem CID | 13367 |
IUPAC Name | prop-2-ene-1-thiol |
SMILES | SCC=C |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
870-23-5 | |
74.14 | |
ULIKDJVNUXNQHS-UHFFFAOYSA-N | |
13367 | |
SCC=C |
C3H6S | |
MFCD00004894 | |
allyl mercaptan, 2-propene-1-thiol, allylthiol, allylmercaptan, allyl sulfhydrate, unii-1x587iby09, 2-propenyl mercaptan, 2-propenyl-1-thiol, ch2=chch2sh, allyl thiol | |
prop-2-ene-1-thiol |
Specifications
870-23-5 | |
0.9300g/mL | |
18°C | |
Glass bottle | |
H2C=CHCH2SH | |
MFCD00004894 | |
0.93 | |
Solubility in water: insoluble. Other solubilities: soluble in alcohol and ether | |
SCC=C | |
74.14 | |
74.13 | |
Technical | |
Allyl mercaptan |
Colorless to Yellow | |
67°C to 68°C | |
Authentic | |
C3H6S | |
1.4745 to 1.4785 | |
01, 440 | |
allyl mercaptan, 2-propene-1-thiol, allylthiol, allylmercaptan, allyl sulfhydrate, unii-1x587iby09, 2-propenyl mercaptan, 2-propenyl-1-thiol, ch2=chch2sh, allyl thiol | |
ULIKDJVNUXNQHS-UHFFFAOYSA-N | |
prop-2-ene-1-thiol | |
13367 | |
70% | |
Liquid |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
May cause respiratory irritation.
Highly flammable liquid and vapor
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 212-792-7
missing translation for 'tsca' : TSCA
RUO – Research Use Only