Learn More
alpha,alpha'-Dibromo-m-xylene, 97%, Thermo Scientific Chemicals
£47.30 - £171.00
Chemical Identifiers
CAS | 626-15-3 |
---|---|
Molecular Formula | C8H8Br2 |
Molecular Weight (g/mol) | 263.95 |
InChI Key | OXHOPZLBSSTTBU-UHFFFAOYSA-N |
Synonym | 1,3-bis bromomethyl benzene, m-xylylene dibromide, alpha,alpha'-dibromo-m-xylene, benzene, 1,3-bis bromomethyl, m-bis bromomethyl benzene, m-xylylene bromide, ccris 1777, m-xylyene dibromide, m-xylene, .alpha.,.alpha.'-dibromo, m-.alpha.,.alpha.'-dibromoxylene |
PubChem CID | 69373 |
IUPAC Name | 1,3-bis(bromomethyl)benzene |
SMILES | C1=CC(=CC(=C1)CBr)CBr |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
11741371
|
Thermo Scientific Acros
155050050 |
5 g | Glass bottle |
£47.30
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11934521
|
Thermo Scientific Acros
155050250 |
25 g | Glass bottle |
£171.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
626-15-3 | |
263.95 | |
1,3-bis bromomethyl benzene, m-xylylene dibromide, alpha,alpha'-dibromo-m-xylene, benzene, 1,3-bis bromomethyl, m-bis bromomethyl benzene, m-xylylene bromide, ccris 1777, m-xylyene dibromide, m-xylene, .alpha.,.alpha.'-dibromo, m-.alpha.,.alpha.'-dibromoxylene | |
1,3-bis(bromomethyl)benzene |
C8H8Br2 | |
OXHOPZLBSSTTBU-UHFFFAOYSA-N | |
69373 | |
C1=CC(=CC(=C1)CBr)CBr |
Specifications
626-15-3 | |
100.0 | |
White to Brown | |
Authentic | |
C8H8Br2 | |
05,374 | |
Solubility in water: insoluble | |
C1=CC(=CC(=C1)CBr)CBr | |
263.95 | |
263.95 | |
Crystalline Powder and/or Chunks |
96.0 | |
75.0°C to 77.0°C | |
135.0°C to 140.0°C (20.0 mmHg) | |
96% min. (HPLC) | |
C6H4(CH2Br)2 | |
1,3-bis bromomethyl benzene, m-xylylene dibromide, alpha,alpha'-dibromo-m-xylene, benzene, 1,3-bis bromomethyl, m-bis bromomethyl benzene, m-xylylene bromide, ccris 1777, m-xylyene dibromide, m-xylene, .alpha.,.alpha.'-dibromo, m-.alpha.,.alpha.'-dibromoxylene | |
OXHOPZLBSSTTBU-UHFFFAOYSA-N | |
1,3-bis(bromomethyl)benzene | |
69373 | |
97% | |
α, α'-Dibromo-m-xylene |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 210-931-6
- alpha, alpha'-Dibromo-m-xylene
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only