missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PCRmax Alpha Cycler 4 Thermal Cycler
PCRmax Alpha Cycler 4 Thermal Cycler has four independently controlled blocks for simplified temperature and space optimization
Brand: PCRmax 93945-10
15231.23 GBP valid until 2024-12-31
Use promo code "21879" to get your promotional price.
This item is not returnable.
View return policy
Description
- These compact thermal cyclers deliver reproducible results every run - with little to no training needed
- Features include a clear responsive touch screen, secure user-specific programming, adjustable heated lid, and active sample cooling for sharper amplification
- A Program Wizard generates a sequence specific protocol in seconds - a quick way to optimize new assays
- Units also retain approximately 1000 reports for reviewing at a later date and run on an intuitive, HD Android™ tablet interface
- Save personal settings to a USB drive; sign in is as simple as inserting your USB drive, no password needed
- The four-block systems take up less space than two or four single-block units and cost less than purchasing multiple units
- Control each block separately for added versatility and use by more than one person at a time, or run on the same program to increase throughput
- Each block also has its own independent heated lid - no interruption of assays running at different settings
Specifications
50°C: ±0.25°C | |
33 x 70 x 53.5 cm | |
100 to 240 VAC | |
HD Android™ Tablet | |
Yes, 35° to 115°C | |
4 Blocks |
10 to 100°C | |
Single 384-well, Triple 96-Well Capacity | |
Thermal Cycler | |
1° to 29°C | |
≤ 1000 Programs |