missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PCRmax Alpha Cycler 4 Thermal Cycler
PCRmax Alpha Cycler 4 Thermal Cycler has four independently controlled blocks for simplified temperature and space optimization
£17845.00 - £17930.00
Specifications
Thermal Accuracy | 50°C: ±0.25°C |
---|---|
Temperature Range (Metric) | 10 to 100°C |
Dimensions | 33 x 70 x 53.5 cm |
Electrical Requirements | 100 to 240 VAC |
Product Type | Thermal Cycler |
Product Code | Brand | Capacity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity | Price | Quantity & Availability | |||||
15569164
|
PCRmax
93945-10 |
Single 384-well, Triple 96-Well Capacity |
£17930.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15522895
|
PCRmax
93945-06 |
Dual 384-well, Dual 96-well Capacity |
£17845.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15512895
|
PCRmax
93945-04 |
Triple 384-well, Single 96-Well Capacity |
£17845.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15532895
|
PCRmax
93945-08 |
Quad 384-well Capacity |
£17930.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- These compact thermal cyclers deliver reproducible results every run - with little to no training needed
- Features include a clear responsive touch screen, secure user-specific programming, adjustable heated lid, and active sample cooling for sharper amplification
- A Program Wizard generates a sequence specific protocol in seconds - a quick way to optimize new assays
- Units also retain approximately 1000 reports for reviewing at a later date and run on an intuitive, HD Android™ tablet interface
- Save personal settings to a USB drive; sign in is as simple as inserting your USB drive, no password needed
- The four-block systems take up less space than two or four single-block units and cost less than purchasing multiple units
- Control each block separately for added versatility and use by more than one person at a time, or run on the same program to increase throughput
- Each block also has its own independent heated lid - no interruption of assays running at different settings
Specifications
50°C: ±0.25°C | |
33 x 70 x 53.5 cm | |
Thermal Cycler | |
1° to 29°C | |
≤ 1000 Programs |
10 to 100°C | |
100 to 240 VAC | |
HD Android™ Tablet | |
Yes, 35° to 115°C |