missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha-D-Galactose pentaacetate, Thermo Scientific Chemicals
£25.13 - £159.00
Chemical Identifiers
CAS | 4163-59-1 |
---|---|
Molecular Formula | C16H22O11 |
Molecular Weight (g/mol) | 390.34 |
MDL Number | MFCD00064081 |
InChI Key | LPTITAGPBXDDGR-UHFFFAOYNA-N |
Synonym | alpha-d-galactose pentaacetate, a-d-galactose pentaacetate, pentaacetyl-alpha-d-galactose, alpha-d-galactose penta-acetate, 1,2,3,4,6-penta-o-acetyl-alpha-d-galactopyranose, alpha-d-galactose-pentaacetate, 1,2,3,4,6-pentaacetyl-alpha-d-galactose, alpha-d-galactose 1,2,3,4,6-pentaacetate, 1,2,3,4,6-penta-o-acetyl-alpha-d-galactose, penta-o-acetyl-alpha-d-galactose |
PubChem CID | 6432426 |
ChEBI | CHEBI:63145 |
IUPAC Name | [(2R,3S,4S,5R,6R)-3,4,5,6-tetraacetyloxyoxan-2-yl]methyl acetate |
SMILES | CC(=O)OCC1OC(OC(C)=O)C(OC(C)=O)C(OC(C)=O)C1OC(C)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15497689
|
Thermo Scientific Alfa Aesar
J66366.06 |
5 g |
£25.13
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15407699
|
Thermo Scientific Alfa Aesar
J66366.14 |
25 g |
£95.07
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15417699
|
Thermo Scientific Alfa Aesar
J66366.18 |
50 g | N/A | ||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
4163-59-1 | |
390.34 | |
LPTITAGPBXDDGR-UHFFFAOYNA-N | |
6432426 | |
[(2R,3S,4S,5R,6R)-3,4,5,6-tetraacetyloxyoxan-2-yl]methyl acetate |
C16H22O11 | |
MFCD00064081 | |
alpha-d-galactose pentaacetate, a-d-galactose pentaacetate, pentaacetyl-alpha-d-galactose, alpha-d-galactose penta-acetate, 1,2,3,4,6-penta-o-acetyl-alpha-d-galactopyranose, alpha-d-galactose-pentaacetate, 1,2,3,4,6-pentaacetyl-alpha-d-galactose, alpha-d-galactose 1,2,3,4,6-pentaacetate, 1,2,3,4,6-penta-o-acetyl-alpha-d-galactose, penta-o-acetyl-alpha-d-galactose | |
CHEBI:63145 | |
CC(=O)OCC1OC(OC(C)=O)C(OC(C)=O)C(OC(C)=O)C1OC(C)=O |
Specifications
4163-59-1 | |
White | |
MFCD00064081 | |
Soluble in methanol at approximately 50mg/ml | |
CC(=O)OCC1OC(OC(C)=O)C(OC(C)=O)C(OC(C)=O)C1OC(C)=O | |
390.34 | |
CHEBI:63145 | |
Powder |
143°C to 144°C | |
C16H22O11 | |
alpha-d-galactose pentaacetate, a-d-galactose pentaacetate, pentaacetyl-alpha-d-galactose, alpha-d-galactose penta-acetate, 1,2,3,4,6-penta-o-acetyl-alpha-d-galactopyranose, alpha-d-galactose-pentaacetate, 1,2,3,4,6-pentaacetyl-alpha-d-galactose, alpha-d-galactose 1,2,3,4,6-pentaacetate, 1,2,3,4,6-penta-o-acetyl-alpha-d-galactose, penta-o-acetyl-alpha-d-galactose | |
LPTITAGPBXDDGR-UHFFFAOYNA-N | |
[(2R,3S,4S,5R,6R)-3,4,5,6-tetraacetyloxyoxan-2-yl]methyl acetate | |
6432426 | |
390.34 | |
alpha-D-Galactose pentaacetate |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only