Learn More
Skytec™ Alpha Sentinel Premium Reusable Full Face Mask
Alpha Sentinel Full Face Masks are ergonomically designed for optimum protection, comfort and PPE compatibility.
Brand:  Skytec™
 View more versions of this product
Product Code. 30242985
  • £208.33 / Each
Estimated Shipment: 08-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • The five-point adjustable head harness removes the feeling of a downward pull, ensuring a balanced, comfortable fit
  • Well-balanced, lightweight (600 g)
  • Available in three sizes for a more tailored fit, increasing respirator/fit factor
  • Raised visor bridge profile protects the visor from scratches when not in use
  • Visor and filter position provides panoramic field of view without any obstructions
  • Low profile shape provides exceptional compatibility with other PPE
  • The Alpha Sentinel filters are fitted with a simple connection which easily clicks into position
  • The large exhalation valve reduces heat and moisture build-up, providing low breathing resistance
  • Easily cleaned and maintained, with spare parts available
  • Tested to EN136:1998 Class 2, ensuring greater robustness and higher flame retardancy
  • Supplied with user seal check covers, which double as particulate filter holder for combination filter
  • Optical insert available
  • Quantitative face fit adapter available
  • The harness is designed to prevent over-rotation and tangling
Specifications
Specifications
CE , UKCA , EN 136:1998 Class 2
Black
Full Facepiece Respirator
Yes
Alpha Sentinel Full Face Mask
Class 2
HEPA, Combination Organic Vapor/Acid Gas/HEPA, Multi-Gas/HEPA
Large
Thermoplastic Elastomer
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Skytec™ Alpha Sentinel Premium Reusable Full Face Mask > Large