Learn More
BD Vacutainer™ Hemogard™ Closure Plastic K3-Edta Tube
Use with whole blood or EDTA plasma sample. BD Vacutainer™ Hemogard™ Closure Plastic K3-EDTA Tube contains the additive K3 Potassium Salt of EDTA that acts as a coagulant.
Brand: BD 367836
Description
- Lavender Hemogard™ Closure
- Used for whole blood or EDTA plasma sample
- The additive K3 Potassium Salt of EDTA is a liquid in the Vacutainer Tube and acts as a coagulant
- Available in sizes 2ml, 3ml and 4ml.
- The difference between K2EDTA and K3EDTA is that the K3 is a liquid solution in the tubes, whereas the K2 is a liquid solution that is spray coated onto the tubes

Specifications
K2 EDTA Tube | |
Plastic | |
2 mL | |
13 mm |
K3 EDTA Additive | |
Hemogard | |
Block | |
ICSH (International Council Society of Haematology) |
Safety and Handling
- BD Vacutainer K3E 3.6 mg Blood Collection Tubes
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
Hazard Statement
- H332-Harmful if inhaled.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P271-Use only outdoors or in a well-ventilated area.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
- MIXTURE LIST-Contains : Tripotassium EDTA Dihydrate
Your input is important to us. Please complete this form to provide feedback related to the content on this product.