Learn More
Ansell™ AlphaTec™ 2000 STANDARD Bound Product Code.: 16884770
Add to basket
Ansell™ AlphaTec™ 2000 STANDARD Bound
Product Code. 16884770
Small
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Product Code. 16884770
Brand: Ansell™ WH20B0015602
Add to basket
Add to basket
AlphaTec™ 2000 STANDARD Bound provides both protection and comfort with exceptional liquid and particulate protection. Ideal for a wide range of industrial applications with low hazard liquid chemical repellence, particle protection, protection from pesticides and barrier to biological agents.
Provides both protection and comfort with exceptional liquid and particulate protection.
  • Ideal for a wide range of industrial applications
  • Low hazard liquid chemical repellence, particle protection, protection from pesticides and barrier to biological agents.
  • Proven protective barrier to low concentration liquid chemicals, liquids, and particulate biological hazards
  • Comfortable to wear featuring moisture vapor permeable fabric to help reduce the risk of heat stress
  • Optimized body fit, improving wearer comfort and safety
  • Anti-static, tested according to EN 1149-5
  • Low linting to help reduce the risk of cross contamination in critical areas
  • Silicone free, critical in spray painting applications
TRUSTED_SUSTAINABILITY
Specifications
Product Type Controlled Environments ESD Coveralls
Size Small
Gender Unisex
Material Microporous Non-woven Polyethylene Laminate
Color White
Disposable Reusable
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Closure Type Zipper
Seam Type Bound
Sterility Non-sterile
Protection Class CE CATEGORY III
Resistance Chemicals, Anti-static, Particle
Collar Type Hood
Certifications/Compliance CE CATEGORY III (Type 5-B, Type 6-B, EN 1073-2, EN 14126, DIN 32781, EN 1149-5)
No. of Pockets None
Product Title
Ansell™ AlphaTec™ 2000 STANDARD Bound >