missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell AlphaTec™ 2000 STANDARD Lab Coat Bound
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
Description
Designed to allow perspiration to escape from the suit and withstand saturation of liquid chemicals.
- Filter 100% of particulates down to 0.01 microns in size
- Proven protective barrier to low concentration liquid chemicals, liquids, and particulate biological hazards
- Comfortable to wear featuring moisture vapor permeable fabric to help reduce the risk of heat stress
- Optimized fit, improving wearer comfort and safety
- Low linting

Specifications
Specifications
Product Type | Controlled Environments Lab Coat |
Size | 2X-Large |
Gender | Unisex |
Material | Microporous Non-woven Polyethylene Laminate |
Color | White |
Closure Type | Zipper |
Cuff Style | Elastic |
Disposable | Reusable |
Resistance | Fluids |
Style | Knee-length |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Special Offers
-
Multiple Manufacturers
Cleanroom Essentials Extravaganza
Gear Up Your Controlled EnvironmentPromo Code: 24151Expires: 2025-05-02
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Ansell AlphaTec™ 2000 STANDARD Lab Coat Bound >
Spot an opportunity for improvement?Share a Content Correction