Learn More
Ansell™ AlphaTec™ 3000 AVANT AIRline Ultrasonically Welded and Taped G05 Product Code.: 17181045
Estimated Shipment: 13-06-2025
to see stock.
Add to basket
Ansell™ AlphaTec™ 3000 AVANT AIRline Ultrasonically Welded and Taped G05
Product Code. 17181045
3X-Large
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Product Code. 17181045
Brand: Ansell™ YE30T0075407G05
Estimated Shipment: 13-06-2025
to see stock.
Add to basket
Add to basket
AlphaTec™ 3000 AVANT AIRline Ultrasonically welded and taped G05 is a fully encapsulated suit designed for use in combination with the AlphaTec™ AVANT STS V.1 airline regulator.
Fully encapsulated suit designed to be used with the permanently attached MICROCHEM™ AVANT STS V.1 airline regulator.
  • Features include air-channel ventilation system
  • Exceptional protection, over 200 chemicals permeation tested, including chemical warfare agents
  • Welded and taped seams, our highest barrier to liquids and particulates
  • Panoramic visor design ensures a good field of vision for the wearer
  • A range of sleeve options available, including double cuffs and permanently attached chemical protective gloves.
  • HEPA filter provides secondary protection from airline contamination and a SMC silencer ensures the noise inside the suit is always below 70 dB (even at maximum airflow)
  • Suit/Belt mounted regulator complete with a low flow warning whistle the AVANT STS regulator can be mounted on the suit and removed for reuse providing cleaning and decontamination is permissible.
  • Emergency rip cord permits rapid doffing of the suit in cases of emergency or undue distress to the wearer
  • Attached socks with elasticated boot overflap - Socks are designed to be worn inside chemical protective boots with the leg overflap worn outside to reduce the potential for chemical ingress
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance CE CATEGORY III (Type 3, EN 1073-1, EN 14594, EN 1149-5)
Color Yellow
For Use With (Application) Transferring liquids and solids between vessels and tanks and process equipment Opening and draining pumps valves or lines Filling, blending and charging of raw materials Mixing or loading chemicals Cleaning of plant and machines Unexpected leakages spills or other releases Loading and unloading trucks and vehicles Product inspection
Hood Type Fully Encapsulated
Material Multi-layer Barrier Laminate Fabric
Size 3X-Large
Ankle Style Attached Sock Boots with Anti-slip PVC Sole
Closure Type Zipper
Cuff Style Double cuffs and permanently attached chemical protective gloves
Disposable Reusable
Gender Unisex
Neck Style Hood
No. of Pockets None
Resistance Chemical I Radioactive I Antistaic
Seam Type ULTRASONICALLY WELDED AND TAPED
Sterility Non-sterile
Product Title
Ansell™ AlphaTec™ 3000 AVANT AIRline Ultrasonically Welded and Taped G05 >