Learn More
Ansell AlphaTec™ 3000 Overboots Ultrasonically Welded Model 406 Product Code.: 17909150
In Stock
to see stock.
Add to basket
Ansell AlphaTec™ 3000 Overboots Ultrasonically Welded Model 406
Product Code. 17909150 Shop All Ansell Products
Size: 42 to 46
Click to view available options
Size: 42 to 46
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 17909150
Brand: Ansell YE30W0040600
In Stock
to see stock.
Add to basket
Add to basket
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
AlphaTec™ 3000 Overboots Ultrasonically Welded Model 406 features a protective multi-layer barrier fabric effective against numerous chemicals.
  • One of the lightest and most comfortable chemical protective garments on the market today, this durable multi-layer fabric provides an extremely effective barrier against both inorganic chemicals and biological hazards.
  • Provides a strong liquid and particle barrier
  • Overboots, elastic to top of boot, tie fastening and welded seams.
TRUSTED_SUSTAINABILITY
Specifications
Size 42 to 46
Material Multi-layer Barrier Laminate Fabric
Color Yellow
Disposable Reusable
Sterility Non-sterile
Seam Type Welded
For Use With (Application) Unexpected leakages spills or other releases, Product inspection, Cleaning of plant and machines, Transferring liquids and solids between vessels and tanks and process equipment, Opening and draining pumps valves or lines, Loading and unloading of trucs and vehicles, Filling, blending and charging of raw materials, Mixing or loading chemicals
Certifications/Compliance CE Number, EN 14126:2003, EN 14605:2005 + A1:2009, EN 14605:2005 + A1:2009, EN 13034:2005 + A1:2009
Resistance Chemical-resistant, Biological Hazards
Product Type Boot Covers
  • Multiple Manufacturers
    Save up to 60% on gloves, apparel, and more
    Promo Code: 24131
    Expires2025-06-08
    New Offer
    Shop Now
Product Content Correction
Product Title
Ansell AlphaTec™ 3000 Overboots Ultrasonically Welded Model 406 >