Learn More
Ansell Microchem™ 4000 Overboots Product Code.: 15586464
Estimated Shipment: 15-05-2025
to see stock.
Add to basket
Ansell Microchem™ 4000 Overboots
Product Code. 15586464 Shop All Ansell Products
Product Code. 15586464
Brand: Ansell GR40W0040600
Estimated Shipment: 15-05-2025
to see stock.
Add to basket
Add to basket
Fits over shoes with EU size 42 to 46 (UK 8-11.) Overboots are constructed from a multi-layer barrier laminate.
  • Double sole design
  • Elasticated opening and tie fastening
  • Ultrasonically welded seams
  • Multi-layer barrier laminate
TRUSTED_SUSTAINABILITY
Specifications
Size Fits over Euro sizes 42-46 (UK 8-11)
Material Multi-layer Barrier Laminate Fabric
Color Green
Closure Type Elastic Opening and Ties
Sole Type Double Sole Design
Certifications/Compliance (Europe) Category I PPE, 89/686/EEC
Packaging Quantity 100/Pk
Product Type Boot Covers
  • Multiple Manufacturers
    Save up to 60% on gloves, apparel, and more
    Promo Code: 24131
    Expires2025-06-08
    New Offer
    Shop Now
Product Content Correction
Product Title
Ansell Microchem™ 4000 Overboots >