missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell AlphaTec™ 4000 AVANT2 AIRline
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Description
- Exceptional protection, over 200 chemicals permeation tested, including chemical warfare agents
- Welded and taped seams, our highest barrier to liquids and particulates.
- Panoramic visor design ensures a good field of vision for the wearer.
- A range of sleeve options available, including double cuffs and permanently attached chemical protective gloves.
- HEPA filter provides secondary protection from airline contamination and a SMC silencer ensures the noise inside the suit is always below 70 dB (even at maximum airflow).
- Emergency rip cord permits rapid doffing of the suit in cases of emergency or undue distress to the wearer.
- Attached socks with elasticated boot overflap - Socks are designed to be worn inside chemical protective boots with the leg overflap worn outside to reduce the potential for chemical ingress.

Specifications
Specifications
Color | Green |
For Use With (Application) | Contaminated Environments, Chemical Handling, Filling, Blending and Charging of Raw Materials, Transferring Liquids and Solids, Opening and Draining Pumps Valves or Lines, Loading and Unloading of Proces Equipment, Raw Material Sample Collection, Concentration Mixing and Palletisation, Hazmat Response, Assembly and Disassembly of Large Pipes |
Material | Multi-layer Barrier Laminate Fabric |
Size | 2X-Large |
Seam Type | Welded and Taped |
Style | G400756 |
Weight (Metric) | 103 gsm |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 6
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Ansell AlphaTec™ 4000 AVANT2 AIRline >
Spot an opportunity for improvement?Share a Content Correction