Learn More
Ansell™ AlphaTec™ 58-530B Series Nitrile Immersion Gloves
GREENER_CHOICE
Enables users to handle wet or oily objects with less grip force and more control. The patented Ansell Grip TechnologyTM ensures that the coating polymer does not penetrate the liner during manufacture significantly reducing the likelihood of chemical leakage onto the skin.
Brand:  Ansell™ 58530B110
49.85 GBP valid until 2024-12-31
Use promo code "21676" to get your promotional price.
 View more versions of this product
Product Code. 12659731
  • £67.95 / 6 pairs
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Nitrile polymer coating reduces chemical leakage onto skin
  • For indoor applications
  • Nylon liner
  • Gauntlet cuff
  • Sea green shell, anthracite grey dip color
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Nitrile
Green, Gray
11
Gauntlet
Black Acrylic
Chemical resistance
Nitrile
Immersion Gloves
58-530B
Acrylic
6 Pairs
58-530B