missing translation for 'onlineSavingsMsg'
Learn More

Tenak™ Alu FlexRack for Chest Freezers

Alu FlexRack is recommended for all applications where a flexible storage solution is needed.

Brand:  Tenak™ TE20110

 View more versions of this product

Product Code. 15280464

  • £96.55 / Each
Estimated Shipment: 22-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Due to the adjustable shelves the rack can be used for all cryobox heights
  • Includes 1 shelf and 1 locking rod for securing the boxes
  • Constructed of aluminum
  • Can be used for any chest freezer
Specifications

Specifications

Locking rod
1
145 mm
140 mm
Aluminum
Chest Frseezers
340 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tenak™ Alu FlexRack for Chest Freezers > Height: 340 mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.