Learn More
Macherey-Nagel™ Alugram Aluminum Sheet CEL 300 UV254
Product Code. 10338651 Shop All Macherey-Nagel Products
Click to view available options
Dimensions (L x W):
5 x 2 cm
20 x 20 cm
Unit Size:
Pack of 25
Pack of 50
Product Code. 10338651
Brand: Macherey-Nagel™ 818156
Only null left
Add to basket
Add to basket
Specifications
Quantity 25 Pk.
Type Aluminum Sheet
Fluorescent Indicator UV254
For Use With (Application) Partition Chromatography Of Polar Substances Such As Amino Acids, Carboxylic Acids Or Carbohydrates
Specific Surface 15,000 cm2/g
Dimensions (L x W) 20 x 20 cm