Learn More
Macherey-Nagel™ Alugram Aluminum Sheet Nano-SIL G/UV254
Brand:  Macherey-Nagel™ 818342
 View more versions of this product
Product Code. 12760813
  • £141.00 / Pack of 50
In Stock
to see stock.
Explore more special offers
Add to basket
Specifications
2 to 10 μm
50 Pk.
UV254
∼500 m2/g
5 x 20 cm
60 Å
Aluminum Sheet
0.75 mL/g
0.20 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Macherey-Nagel™ Alugram Aluminum Sheet Nano-SIL G/UV254 > Plate Size: 50 x 200mm